1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene ≥97%
Synonyms:
(S,S)-Isopropyl-DuPhos
H60407-MD
H60407-03
H60407-MD
H60407-03
AAH60407-MDEA
180.59
USD
AAH60407-MD
CAAAH60407-03
CAAAH60407-MD
AAH60407-03
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene ≥97%
1,2-Bis[(2S,5S)-2,5-diisopropyl-1-phospholanyl]benzene
Formula:
C₂₆H₄₄P₂ MW: 418.58 g/mol Melting Pt: 26…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684550 CAS Number: 147253-69-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...