(S)-2-Diphenylphosphino-1-phenylethylamine ≥97%

Warning

Synonyms: (S)-2-(Diphenylphosphino)-1-phenylethanamine

H60404-MD H60404-03 H60404-MD H60404-03
AAH60404-03EA 300.24 USD
CAAAH60404-03 AAH60404-03 AAH60404-MD CAAAH60404-MD
(S)-2-Diphenylphosphino-1-phenylethylamine ≥97%
(S)-2-Diphenylphosphino-1-phenylethylamine
Formula: C₂₀H₂₀NP
MW: 305.361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17013989
CAS Number: 1103533-85-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR