(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) 97%
|
Danger
|
Synonyms:
(R)-(-)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulfonate)
H60401-03
H60401-03
AAH60401-03EA
106.27
USD
CAAAH60401-03
AAH60401-03
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) 97%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate)
Formula:
C₂₂H₁₂F₆O₆S₂ MW: 550.44 g/mol Melting Pt: 82…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274615 CAS Number: 126613-06-7 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...