4-O-Benzyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal 97%

H60280-MD H60280-03 H60280-03 H60280-MD
AAH60280-MDEA 45.62 USD
AAH60280-MD AAH60280-03 CAAAH60280-MD CAAAH60280-03
4-O-Benzyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal 97%
4-O-Benzyl-3,6-di-O-tert-butyldimethylsilyl-D-glucal
Formula: C₂₅H₄₄O₄Si₂
MW: 464.793 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22989001
CAS Number: 111830-58-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR