Bis[2-(diphenylphosphino)ethyl]ammonium chloride ≥97%

Warning

Synonyms: Bis(2-(diphenylphosphino)ethyl)amine hydrochloride

H60250-06 H60250-06 H60250-MD H60250-MD H60250-03 H60250-03
AAH60250-03EA 133.95 USD
CAAAH60250-06 CAAAH60250-MD AAH60250-03 AAH60250-06 AAH60250-MD CAAAH60250-03
Bis[2-(diphenylphosphino)ethyl]ammonium chloride ≥97%
Bis[2-(diphenylphosphino)ethyl]ammonium chloride
Formula: C₂₈H₃₀ClNP₂
MW: 477.953 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00061569
CAS Number: 66534-97-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR