1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene ≥97%

Synonyms: 1,2-Bis((2R,5R)-2,5-diisopropylphospholan-1-yl)benzene(+)-1,2-Bis((2R,5R)-2,5-di-i-propylphospholano)benzene(R,R)-i-Pr-DUPHOS

H60227-03 H60227-MD H60227-03 H60227-MD
AAH60227-MDEA 194.68 USD
CAAAH60227-MD AAH60227-MD AAH60227-03 CAAAH60227-03
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene ≥97%
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]benzene
Formula: C₂₆H₄₄P₂
MW: 418.586 g/mol
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02684549
CAS Number: 136705-65-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR