1-(2-Bromophenylsulfonyl)-3-methylpiperidine 97%
|
Warning
|
Synonyms:
1-(2-Bromophenylsulfonyl)-3-methylpiperidine
H60206-MD
H60206-03
H60206-MD
H60206-03
AAH60206-03EA
194.25
USD
CAAAH60206-MD
CAAAH60206-03
AAH60206-03
AAH60206-MD
1-(2-Bromophenylsulfonyl)-3-methylpiperidine 97%
1-(2-Bromophenylsulfonyl)-3-methylpiperidine
Formula:
C₁₂H₁₆BrNO₂S MW: 318.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12650340 CAS Number: 1184609-55-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...