Methyl-2-bromohexanoate 98%

Warning

Synonyms: Methyl 2-bromohexanoate

H60165-AE H60165-AC H60165-AC H60165-AE
AAH60165-ACEA 32.13 USD
CAAAH60165-AE AAH60165-AC AAH60165-AE CAAAH60165-AC
Methyl-2-bromohexanoate 98%
Methyl 2-bromohexanoate
Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
Density: 1.289
Flash Pt: 69 °C(156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045038
EINECS: 226-643-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR