Asiatic acid 97%

Supplier: Thermo Scientific Chemicals

H60149-ME H60149-ME
AAH60149-MEEA 276.99 USD
AAH60149-ME CAAAH60149-ME
Asiatic acid 97%
Asiatic acid
Formula: C₃₀H₄₈O₅
Melting Pt: 325…330 °C
Storage Temperature: Ambient
MDL Number: MFCD00238541
CAS Number: 464-92-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR