trans-1,4-Cyclohexanedicarboxylic acid 95%
|
Warning
|
H60089-06
H60089-03
H60089-03
H60089-06
AAH60089-03EA
44.34
USD
CAAAH60089-06
AAH60089-03
CAAAH60089-03
AAH60089-06
trans-1,4-Cyclohexanedicarboxylic acid 95%
trans-1,4-Cyclohexanedicarboxylic acid
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066203 CAS Number: 619-82-9 EINECS: 210-614-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...