(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-tert-Butyl 2-(hydroxymethyl)morpholine-4-carboxylate
, (R)-N-Boc-2-hydroxymethylmorpholine
H60062-03
H60062-03
AAH60062-03EA
151.99
USD
CAAAH60062-03
AAH60062-03
(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 99%
(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate
Formula:
C10H19NO4 MW: 217.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09260605 CAS Number: 135065-71-3 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...