2-Amino-3-chloro-1,4-naphthoquinone 97%

Danger

H60040-MC H60040-MC
AAH60040-MCEA 363.15 USD
AAH60040-MC CAAAH60040-MC
2-Amino-3-chloro-1,4-naphthoquinone 97%
2-Amino-3-chloro-1,4-naphthoquinone
Formula: C₁₀H₆ClNO₂
MW: 207.61 g/mol
Melting Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00001680
CAS Number: 2797-51-5
EINECS: 220-529-2
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR