N-(3,5-Dichlorophenyl)benzenesulfonamide 97%

Warning

H60031-03 H60031-MD H60031-03 H60031-MD
AAH60031-MDEA 194.67 USD
CAAAH60031-03 AAH60031-MD AAH60031-03 CAAAH60031-MD
N-(3,5-Dichlorophenyl)benzenesulfonamide 97%
N-(3,5-Dichlorophenyl)benzenesulfonamide
Formula: C₁₂H₉Cl₂NO₂S
MW: 302.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01212617
CAS Number: 54129-15-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR