1-Methyl-3-octylimidazolium hexafluorophosphate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59990-14
H59990-14
H59990-06
H59990-06
AAH59990-14EA
252.16
USD
CAAAH59990-14
AAH59990-14
AAH59990-06
CAAAH59990-06
1-Methyl-3-octylimidazolium hexafluorophosphate 95%
1-Methyl-3-octylimidazolium hexafluorophosphate
Formula:
C₁₂H₂₃F₆N₂P MW: 340.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06798203 CAS Number: 304680-36-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...