1-Bromo-4,5-dichloro-2-fluorobenzene 96%
|
Warning
|
Synonyms:
4,5-Dichloro-2-fluorobromobenzene
H59984-03
H59984-03
H59984-MD
H59984-MD
AAH59984-MDEA
117.42
USD
AAH59984-MD
CAAAH59984-MD
CAAAH59984-03
AAH59984-03
1-Bromo-4,5-dichloro-2-fluorobenzene 96%
1-Bromo-4,5-dichloro-2-fluorobenzene
Formula:
C₆H₂BrCl₂F MW: 243.884 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878158 CAS Number: 1000572-78-0 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...