N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-L-tyrosineFmoc-Tyr-OH

H59967-03 H59967-03
AAH59967-03EA 64.99 USD
CAAAH59967-03 AAH59967-03
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine
Formula: C₂₄H₂₁NO₅
MW: 403.43 g/mol
Melting Pt: 182…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00134890
CAS Number: 92954-90-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR