1-(3-Chlorophenyl)piperazine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H59896-06 H59896-14 H59896-14 H59896-06
AAH59896-14EA 163.99 USD
CAAAH59896-14 AAH59896-14 CAAAH59896-06 AAH59896-06
1-(3-Chlorophenyl)piperazine hydrochloride 97%
1-(3-Chlorophenyl)piperazine hydrochloride
Formula: C₁₀H₁₃ClN₂·xHCl
MW: 196.68 g/mol (anhy)
Melting Pt: 210…214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00039032
CAS Number: 65369-76-8
EINECS: 265-718-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR