Z-Hyp-OH 99%

Synonyms: trans-N-Cbz-4-hydroxy-L-prolineN-Cbz-Hydroxy-L-ProlineZ-L-4-Hydroxyproline

H59882-14 H59882-06 H59882-06 H59882-14
AAH59882-14EA 202.89 USD
CAAAH59882-14 CAAAH59882-06 AAH59882-14 AAH59882-06
Z-Hyp-OH 99%
Z-Hyp-OH
Formula: C13H15NO5
MW: 265.3 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00037329
CAS Number: 13504-85-3
EINECS: 236-831-2

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR