N-Ethyl-3-methylbenzamide 97%

Warning

H59871-MD H59871-03 H59871-MD H59871-03
AAH59871-MDEA 156.56 USD
CAAAH59871-03 CAAAH59871-MD AAH59871-MD AAH59871-03
N-Ethyl-3-methylbenzamide 97%
N-Ethyl-3-methylbenzamide
Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00496601
CAS Number: 26819-07-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR