1-Benzyl-3-methylimidazolium tetrafluoroborate 97%

Warning

Synonyms: 1-Benzyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

H59732-18 H59732-18 H59732-06 H59732-06
AAH59732-06EA 42.23 USD
AAH59732-06 CAAAH59732-18 AAH59732-18 CAAAH59732-06
1-Benzyl-3-methylimidazolium tetrafluoroborate 97%
1-Benzyl-3-methylimidazolium tetrafluoroborate
Formula: C₁₁H₁₃BF₄N₂
MW: 260.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784449
CAS Number: 500996-04-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR