1-Ethyl-2,3-dimethylimidazoliumtrifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

H59726-18 H59726-18 H59726-06 H59726-06
AAH59726-06EA 89.69 USD
CAAAH59726-18 CAAAH59726-06 AAH59726-06 AAH59726-18
1-Ethyl-2,3-dimethylimidazoliumtrifluoromethanesulfonate 98%
1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulphonate
Formula: C₈H₁₃F₃N₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD07784438
CAS Number: 174899-72-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR