4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester 96%

H59683-MD H59683-03 H59683-03 H59683-MD
AAH59683-03EA 497.49 USD
CAAAH59683-MD AAH59683-03 AAH59683-MD CAAAH59683-03
4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester 96%
4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester
Formula: C₁₂H₁₄BCl₂FO₂
MW: 290.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660310
CAS Number: 1116681-96-9

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR