Isobutyl propionate 98%
|
Warning
|
Synonyms:
Propionic acid Isobutyl ester
H59612-AC
H59612-AC
H59612-AE
H59612-AE
AAH59612-AEEA
66.15
USD
AAH59612-AE
AAH59612-AC
CAAAH59612-AE
CAAAH59612-AC
Isobutyl propionate 98%
Isobutyl propionate
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Melting Pt: –71 °C Density: 0.869 Flash Pt: 66 °C(151 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009307 CAS Number: 540-42-1 EINECS: 208-746-0 UN: 2394 ADR: "3",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...