1-Ethyl-2,3-dimethylimidazoliumtetrafluoroborate 98%

Warning

Synonyms: 1-ethyl-2,3-dimethylimidazolium tetrafluoroborate

H59541-18 H59541-06 H59541-06 H59541-18
AAH59541-06EA 34.76 USD
AAH59541-06 AAH59541-18 CAAAH59541-18 CAAAH59541-06
1-Ethyl-2,3-dimethylimidazoliumtetrafluoroborate 98%
1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate
Formula: C₇H₁₃BF₄N₂
MW: 212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458929
CAS Number: 307492-75-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR