tert-Butyl methyl malonate 95%
Warning
|
Synonyms:
tert.-Butyl methyl malonate
H59540-9W
H59540-9W
H59540-AB
H59540-AB
AAH59540-ABEA
166.99
USD
AAH59540-AB
CAAAH59540-9W
CAAAH59540-AB
AAH59540-9W
tert-Butyl methyl malonate 95%
tert-Butyl methyl malonate
Formula:
C₈H₁₄O₄ MW: 174.19 g/mol Boiling Pt: 80 °C/11 mm Density: 1.030 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042856 EINECS: 255-919-1 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...