2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate 98%

Warning

Synonyms: CIP

H59515-06 H59515-03 H59515-03 H59515-06
AAH59515-03EA 50.87 USD
AAH59515-03 CAAAH59515-06 AAH59515-06 CAAAH59515-03
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate 98%
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate
Formula: C₅H₁₀ClF₆N₂P
MW: 278.56 g/mol
Melting Pt: 231…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00191914
CAS Number: 101385-69-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR