1-Ethyl-3-methylimidazoliumthiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate
H59493-06
H59493-06
H59493-18
H59493-18
AAH59493-18EA
591.63
USD
CAAAH59493-18
CAAAH59493-06
AAH59493-18
AAH59493-06
1-Ethyl-3-methylimidazoliumthiocyanate 98%
1-Ethyl-3-methylimidazolium thiocyanate
Formula:
C₇H₁₁N₃S MW: 169.25 g/mol Melting Pt: –6 °C Density: 1.14 Storage Temperature: Ambient |
MDL Number:
MFCD06798182 CAS Number: 331717-63-6 UN: 2810 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...