Ethyl-4,6-dichloro-8-methylquinoline-3-carboxylate 97%

Warning

H59474-MD H59474-03 H59474-MD H59474-03
AAH59474-03EA 126.69 USD
CAAAH59474-03 AAH59474-03 CAAAH59474-MD AAH59474-MD
Ethyl-4,6-dichloro-8-methylquinoline-3-carboxylate 97%
Ethyl-4,6-dichloro-8-methylquinoline-3-carboxylate
Formula: C₁₃H₁₁Cl₂NO₂
MW: 284.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173360
CAS Number: 338954-50-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR