4-Chloro-6,8-difluoroquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

H59428-03 H59428-MD H59428-03 H59428-MD
AAH59428-MDEA 64.99 USD
CAAAH59428-03 AAH59428-MD AAH59428-03 CAAAH59428-MD
4-Chloro-6,8-difluoroquinoline 97%
4-Chloro-6,8-difluoroquinoline
Formula: C₉H₄ClF₂N
MW: 199.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00278782
CAS Number: 239463-89-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR