N-benzyl-4-methoxybenzamide 97%

Warning

H59421-MD H59421-MD H59421-03 H59421-03
AAH59421-03EA 462.87 USD
AAH59421-03 AAH59421-MD CAAAH59421-03 CAAAH59421-MD
N-benzyl-4-methoxybenzamide 97%
N-benzyl-4-methoxybenzamide
Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00507374
CAS Number: 7465-87-4

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR