Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

4-Chloro-N,N-diisopropylbutyramide ≥97%

Supplier: Alfa Aesar
Synonyms: 4-Chloro-N,N-diisopropylbutyramide

H59335-03 H59335-MD H59335-MD H59335-03
AAH59335-MDEA 141.11 USD InStock
CAAAH59335-03 AAH59335-MD CAAAH59335-MD AAH59335-03
4-Chloro-N,N-diisopropylbutyramide ≥97%
4-Chloro-N,N-diisopropylbutyramide


Formula: C₁₀H₂₀ClNO
MW: 205.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683057
CAS Number: 4635-60-3
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty