4,6-Dichloroquinoline 97%
|
Warning
|
H59307-MD
H59307-03
H59307-MD
H59307-03
AAH59307-MDEA
56.86
USD
CAAAH59307-03
CAAAH59307-MD
AAH59307-MD
AAH59307-03
4,6-Dichloroquinoline 97%
4,6-Dichloroquinoline
Formula:
C₉H₅Cl₂N MW: 198.05 g/mol Melting Pt: 104…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156141 CAS Number: 4203-18-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...