4-Chloro-N-(2-iodophenyl)benzamide 97%

Warning

H59260-MD H59260-03 H59260-MD H59260-03
AAH59260-03EA 185.37 USD
CAAAH59260-MD AAH59260-03 AAH59260-MD CAAAH59260-03
4-Chloro-N-(2-iodophenyl)benzamide 97%
4-Chloro-N-(2-iodophenyl)benzamide
Formula: C₁₃H₉ClINO
Storage Temperature: Ambient
MDL Number: MFCD00783636
CAS Number: 346689-44-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR