5-Bromo-2-hydroxy-6-picoline 97%

Danger

Synonyms: 3-Bromo-6-hydroxy-2-methylpyridine5-Bromo-6-methylpyridin-2-ol

H59227-03 H59227-03
AAH59227-03EA 94.46 USD
AAH59227-03 CAAAH59227-03
5-Bromo-2-hydroxy-6-picoline 97%
5-Bromo-2-hydroxy-6-picoline
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD03427653
CAS Number: 54923-31-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR