Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2,4-dimethoxyaniline 98%

Supplier: Alfa Aesar
Warning

H59214-03 H59214-03
AAH59214-03EA 30.59 USD InStock
AAH59214-03 CAAAH59214-03
5-Chloro-2,4-dimethoxyaniline 98%
5-Chloro-2,4-dimethoxyaniline


Formula: C₈H₁₀ClNO₂
MW: 187.63 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 87-90°C
Storage Temperature: Ambient
MDL Number: MFCD00025752
CAS Number: 97-50-7
EINECS: 202-586-5
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty