Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester ≥96%

Supplier: Alfa Aesar

H59212-03 H59212-MD H59212-MD H59212-03
AAH59212-03EA 313.12 USD InStock
AAH59212-03 AAH59212-MD CAAAH59212-MD CAAAH59212-03
2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester ≥96%
2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester


Formula: C₁₂H₁₆BFO₃
MW: 238.065 g/mol
Melting Pt: 110-112°C
Storage Temperature: Ambient
MDL Number: MFCD16994421
CAS Number: 1029439-02-8
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty