1-Hexyl-3-methylimidazolium trifluoromethanesulfonate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Hexyl-3-methylimidazolium Trifluoromethanesulfonate1-Hexyl-3-methylimidazolium triflate1-Hexyl-3-methylimidazolium Trifluoromethanesulphonate

H59210-14 H59210-06 H59210-14 H59210-06
AAH59210-14EA 363.16 USD
CAAAH59210-14 AAH59210-14 CAAAH59210-06 AAH59210-06
1-Hexyl-3-methylimidazolium trifluoromethanesulfonate 95%
1-Hexyl-3-methylimidazolium trifluoromethanesulphonate
Formula: C₁₁H₁₉F₃N₂O₃S
MW: 316.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427621
CAS Number: 460345-16-8

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR