Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Difluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester ≥96%

Supplier: Alfa Aesar
Synonyms: 2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester

H59195-03 H59195-03 H59195-MD H59195-MD
AAH59195-03EA 372.86 USD InStock
CAAAH59195-03 AAH59195-03 AAH59195-MD CAAAH59195-MD
2,6-Difluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester ≥96%
2,6-Difluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester


Formula: C₁₄H₁₇BF₂O₄
MW: 298,09
Storage Temperature: Ambient
MDL Number: MFCD12546491
CAS Number: 1218791-32-2
EINECS: 000-000-0
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty