4-(Trifluoromethyl)-1-indanone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59165-06
H59165-03
H59165-03
H59165-06
AAH59165-06EA
340.77
USD
CAAAH59165-03
AAH59165-06
AAH59165-03
CAAAH59165-06
4-(Trifluoromethyl)-1-indanone 97%
4-(Trifluoromethyl)-1-indanone
Formula:
C₁₀H₇F₃O MW: 200.16 g/mol Boiling Pt: 67…72 °C/0.1 mm Melting Pt: 38…42 °C Flash Pt: >108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD07772121 CAS Number: 68755-42-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...