Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine 95%

Synonyms: Nalpha-Fmoc-L-glutamineFmoc-Gln-OH

H59159-03 H59159-06 H59159-03 H59159-06
AAH59159-06EA 74.28 USD
AAH59159-06 CAAAH59159-03 CAAAH59159-06 AAH59159-03
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine 95%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine
Formula: C₂₀H₂₀N₂O₅
MW: 368.39 g/mol
Melting Pt: 224 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037137
CAS Number: 71989-20-3
EINECS: 276-254-3

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR