6,8-Dibromoimidazo[1,2-a]pyrazine 95%

Supplier: Thermo Scientific Chemicals
Warning

H59151-03 H59151-06 H59151-06 H59151-03
AAH59151-06EA 308.99 USD
AAH59151-06 CAAAH59151-06 AAH59151-03 CAAAH59151-03
6,8-Dibromoimidazo[1,2-a]pyrazine 95%
6,8-Dibromoimidazo[1,2-a]pyrazine
Formula: C6H3Br2N3
MW: 276.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460056
CAS Number: 63744-22-9

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR