2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid pinacol ester 96%

Warning

Synonyms: Methyl 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

H59147-03 H59147-MD H59147-03 H59147-MD
AAH59147-03EA 341.96 USD
AAH59147-03 AAH59147-MD CAAAH59147-03 CAAAH59147-MD
2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid pinacol ester 96%
2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
Formula: C₁₄H₁₈BFO₄
MW: 280.102 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD12923177
CAS Number: 757982-31-3

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR