6-Aminoquinoline-5-carbonitrile 95%

Warning

H59061-MD H59061-03 H59061-03 H59061-MD
AAH59061-03EA 109.18 USD
CAAAH59061-MD AAH59061-03 CAAAH59061-03 AAH59061-MD
6-Aminoquinoline-5-carbonitrile 95%
6-Aminoquinoline-5-carbonitrile
Formula: C₁₀H₇N₃
MW: 169.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05855932
CAS Number: 54398-51-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR