N-(4-Chlorobenzyl)-3,5-dinitrobenzamide 97%

Warning

H58730-ME H58730-ME H58730-MC H58730-MC
AAH58730-MCEA 88.17 USD
CAAAH58730-ME CAAAH58730-MC AAH58730-MC AAH58730-ME
N-(4-Chlorobenzyl)-3,5-dinitrobenzamide 97%
N-(4-Chlorobenzyl)-3,5-dinitrobenzamide
Formula: C₁₄H₁₀ClN₃O₅
Storage Temperature: Ambient
MDL Number: MFCD00757362
CAS Number: 446855-41-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR