Phenylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

ChemSeal
H58659-AD H58659-AD
AAH58659-ADEA 261.26 USD
CAAAH58659-AD AAH58659-AD
Phenylzinc bromide 0.5 M in THF
Phenylzinc bromide
Formula: C₆H₅BrZn
MW: 222.39 g/mol
Density: 0.99
Storage Temperature: Ambient
MDL Number: MFCD01319861
CAS Number: 38111-44-3
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR