Isopropylzinc bromide 0.5 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Propylzinc bromide
Packaged under Argon in resealable ChemSeal™ bottles.
ChemSeal
H58536-AD
H58536-AD
AAH58536-ADEA
237.99
USD
CAAAH58536-AD
AAH58536-AD
Isopropylzinc bromide 0.5 M in THF
Isopropylzinc bromide
Isopropylzinc bromide is employed in croos-coupling reactions.
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and water.
Formula:
C₃H₇BrZn MW: 188.373 g/mol Density: 0.963 Storage Temperature: Ambient |
MDL Number:
MFCD06200639 CAS Number: 77047-87-1 UN: 3399 ADR: "4.3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...