Cyclopropylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

ChemSeal
H58008-AD H58008-AD
AAH58008-ADEA 261.99 USD
AAH58008-AD CAAAH58008-AD
Cyclopropylzinc bromide 0.5 M in THF
Cyclopropylzinc bromide
Formula: C₃H₅BrZn
Density: 0.969
Storage Temperature: Ambient
MDL Number: MFCD08741505
CAS Number: 126403-68-7
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR