Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide ≥97%

Supplier: Alfa Aesar
Warning

H57808-03 H57808-MD H57808-03 H57808-MD
AAH57808-03EA 275.01 USD InStock
AAH57808-03 CAAAH57808-MD CAAAH57808-03 AAH57808-MD
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide ≥97%
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide


Formula: C₁₆H₁₉NO₃S
MW: 305.392 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02007969
CAS Number: 349085-82-1
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty