1-Boc-3-methylazetidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl 3-ethynylazetidine-1-carboxylate
H57496-ME
H57496-ME
H57496-MD
H57496-MD
AAH57496-MEEA
335.99
USD
CAAAH57496-MD
CAAAH57496-ME
AAH57496-ME
AAH57496-MD
1-Boc-3-methylazetidine 95%
1-Boc-3-methylazetidine
Formula:
C₉H₁₇NO₂ MW: 171.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09951813 CAS Number: 1314984-00-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...