N-(3,5-Difluorophenyl)-4-methylbenzamide 97%

Warning

H57145-MD H57145-03 H57145-MD H57145-03
AAH57145-MDEA 82.71 USD
AAH57145-MD AAH57145-03 CAAAH57145-MD CAAAH57145-03
N-(3,5-Difluorophenyl)-4-methylbenzamide 97%
N-(3,5-Difluorophenyl)-4-methylbenzamide
Formula: C₁₄H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD07081986
CAS Number: 881763-65-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR